CONSTIX®, Sealed Polyester Swabs, SP-6 Series, Contec®
Supplier: Contec
SP-6
76057-730CS 147.54 USD
76057-730
CONSTIX®, Sealed Polyester Swabs, SP-6 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SP-6 sealed polyester swabs consist of 100% knit polyester fabric that is thermally bonded to a flex-tip polypropylene handle.
  • Flexible tip provides a softer application on surfaces
  • Polyester head provides enhanced sorbency, particle entrapment, and removal
  • 100% polyester headis compatible with most common solvents, such as alcohols and ketones
The swab head is made of cleanroom laundered, double-layer knitted polyester. The polyester head is constructed without adhesives or binders. The swab is durable, absorbent, and features extremely lowMore Product Information