CONSTIX®, Sealed Polyester Swabs, SP-11 Series, Contec®
Supplier: Contec
SP-11
76057-728CS 85.24 USD
76057-728
CONSTIX®, Sealed Polyester Swabs, SP-11 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SP-11 sealed polyester swabs consist of 100% knitted polyester fabric that is thermally bonded to a rigid polypropylene handle.
  • Polyester head provides enhanced sorbency, particle entrapment, and removal
  • Polyester head is compatible with most common solvents, such as alcohols and ketones
  • Flexible tip enables access to very small areas and components
The swab head is made of cleanroom laundered, double-layer knitted microfiber polyester material. The polyester head is thermally sealed and attached, constructed without adhesives or binders. The polypropylene handle has a thin tip. The swab is durable, absorbent, and features extremely low particles and residue levels.