CONSTIX®, Sealed Polyester Swabs, SP-8 Series, Contec®
Supplier: Contec
SP-8
76057-704CS 50.71 USD
76057-704
CONSTIX®, Sealed Polyester Swabs, SP-8 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SP-8 sealed polyester swabs consist of 100% knitted polyester fabric that is thermally bonded to a polypropylene handle with a large paddle.
  • Polyester head provides enhanced sorbency, particle entrapment, and removal
  • Large paddle swab head provides broader surface coverage in critical applications
The swab head is made of cleanroom laundered, double-layer knitted microfiber polyester material. The polyester head is constructed without adhesives or binders, which makes the swab compatible with most common solvents, such as alcohols and ketones. The swab is durable, absorbent, and features extremely low particles and residue levels.