VWR® Upright Freezer Drawer Rack for 96-Well Microtube Boxes
Supplier: VWR International
76051-748
76051-748EA 129.34 USD
76051-748
VWR® Upright Freezer Drawer Rack for 96-Well Microtube Boxes
Racks Cryogenic Box Racks
Designed for use with 96-well microtube boxes.
  • Made of corrosion resistant stainless steel
  • Security lock devices are available for each rack
Drawer racks allow easy access to fewer microtiter plates at a time thus protecting other samples in the rack from warming up.