First Aid Burn Cream, 12 Per Box, First Aid Only
Supplier: First Aid Only
13-006-091
76048-932BX 4.79 USD
76048-932
First Aid Burn Cream, 12 Per Box, First Aid Only
First Aid Supplies Burn Care Supplies
First aid burn cream is perfect for minor cuts, burns, and abrasions.
  • Unit dose packages provide quick and effective application
  • Active ingredient: lidocaine HCL 0.5%, benzalkonium chloride 0.13%
This antiseptic and external analgesic helps prevent infection, relieves pain, and provides temporary relief.