Reagent alcohol 190° Proof

Supplier: Decon Labs
Danger

12R1901
76048-924CS 244.74 USD
76048-924
Reagent alcohol 190° Proof
Alcohol

Reagent alcohol 190 proof is a specially denatured ethanol product used in the clinical histology lab.


  • Denatured according to formula 3A


Appearance: Clear
Odor: Mild

Boiling Pt: 78 °C
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR