BDD, Bacdown Detergent Disinfectant, Concentrated Liquid, Decon Labs, Inc.
Supplier: Decon Labs
8004
76048-922CS 337.88 USD
76048-922
BDD, Bacdown Detergent Disinfectant, Concentrated Liquid, Decon Labs, Inc.
Cleaning Agents Disinfectants
BDD™ (Bacdown® Detergent Disinfectant) will safely and effectively clean and disinfect surfaces in labs, production areas and process equipment.  Diluted two (2) ounces per gallon of water (1:64), BDD™ is a bactericidal, virucidal, fungicidal disinfectant, effective against HIV-1, HBV and more. 
  • Concentrated quaternary (quat) disinfectant
  • Phosphate-free
Certifications: Conforms to OSHA's Bloodborne Pathogens Standard. EPA registered.