F.I.T.™ Comfort Deck Open, Wearwell®
Supplier: Wearwell
548.58X12X12BK-CS20
76048-788EA 10.48 USD
76048-788 76048-790
F.I.T.™ Comfort Deck Open, Wearwell®
Floor Mats Traction and Anti-Fatigue Floor Mats
Most versatile modular tile.
  • Customizable in 3" increments
  • Secure and seamless connections
  • Ability to replace single tiles saves money and down time
  • Silicone-free
The most versatile 12×12" interlocking ergonomic flooring system in the market. The patented LockSafe® connections simply snap together and configure to fit your exact workspace. The top surface and underside of F.I.T. tiles are designed to allow rapid drainage of fluids.