Biohazard Authorized Personnel Only Sign, Brady Worldwide
Supplier: Brady Worldwide
62812
76046-534EA 49.78 USD
76046-534
Biohazard Authorized Personnel Only Sign, Brady Worldwide
Signs
Chemical, Biohazard, and Hazardous Material signs.
  • Graphics are permanently embedded within the sign panel, allowing for use in the most severe environments
  • Excellent chemical and abrasion resistance
  • Ideal for both indoor and outdoor applications
  • Service temperature of –40 to 90 °C (–40 to 193 °F)
Extremely durable B-120 Premium Fiberglass will not chip, fade, rust, shatter, or peel for up to 25 years.