Cole-Parmer® SHM-250 Series Digi-Mantle Heating Kits, Antylia Scientific
Supplier: Antyila Scientific
36204-21 36204-27 36204-17 36204-25 36204-11
47742-566EA 6860.9 USD
47742-566 76045-874 76045-876 76045-878 76045-880
Cole-Parmer® SHM-250 Series Digi-Mantle Heating Kits, Antylia Scientific
Heaters Heating Mantles
Heating kit includes a heating mantle controller and interchangeable heating cartridge.
  • Achieve uniform temperature up to 450 °C across round bottom flask
  • Stirring speeds from 100 to 2000 rpm with excellent stability
  • Vented casing keeps heating cartridge cool to the touch
  • Modular design allows different size heating cartridges to be used on the same Digi-Mantle controller
Control temperature and stirring speed independently on the Digi-Mantle digital controller. The control panel features indicator lights for power, heater, and stirrer. Microprocessor-controlled heater displays the heat setting as a percentage of the total power. View the actual temperature (up to 220 °C) of the flask content on a three digit LCD display by using the included temperature probe.
Each digi-mantle heating kit includes a controller and a heating cartridge that accommodates 100 to 250 ml, 500 or 1000 ml round bottom flasks. Due to its modular design a different size heating cartridge can be used with the same controller, saving money.
Ordering information: Supplied with a support rod. Clamps are not supplied.