Human Recombinant Growth and differentiation factor-15
Supplier: Enzo Life Sciences
201-803-0005
76045-524EA 206.01 USD
76045-524 76045-526 76288-306
Human Recombinant Growth and differentiation factor-15
Proteins and Peptides
Produced in E. coli. Non-glycosylated, disulfide linked homodimer, containing two identical 113 amino acid chains.
  • Carrier-free