Carbol fuchsin (Kinyoun’s) stain for bacteriology, for AFB (acid fast bacteria)

Supplier: HARDY DIAGNOSTICS

Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

CF008
76045-454EA 26.99 USD
76045-454
Carbol fuchsin (Kinyoun’s) stain for bacteriology, for AFB (acid fast bacteria)
Carbol fuchsin

Carbol Fuchsin, Kinyoun for AFB stain, for Mycobacteria


An aqueous solution of basic fuchsin, phenol, and reagent alcohol for the “Cold Stain” method.

Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
MDL Number: MFCD00013129
CAS Number: 4197-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR