FLIPR Membrane Potential Assay Kit, Molecular Devices
Supplier: Molecular Devices
R8128
76044-680EA 846.22 USD
76044-680
FLIPR Membrane Potential Assay Kit, Molecular Devices
Assays Cellular Assays
The FLIPR Membrane Potential Assay Kit includes homogenous florescence-based formulations for observation of real time membrane potential changes.
Delivery information: The FLIPR Membrane Assay Kit includes: Membrane Potential Assay BLUE (five vials), Membrane Potential Assay RED (five vials), one bottle 1X Assay Buffer, and 1X Hanks BSS with 20 mM HEPES, pH 7.4 with 96-well plate.