Anti-IgG Donkey Polyclonal Antibody (Biotin)

Supplier: Tonbo Biosciences
30-8080-M001
76040-392EA 211.2 USD
76040-392
Anti-IgG Donkey Polyclonal Antibody (Biotin)
Antibodies
Donkey anti-Goat IgG (H&L) - Affinity Pure, Biotin Conjugate, min x w/human, mouse, rabbit or rat IgG

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Donkey
Isotype:
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR