Anti-IgG Goat Polyclonal Antibody (Biotin)

Supplier: Tonbo Biosciences
30-8054-U500
76040-324EA 183.7 USD
76040-324
Anti-IgG Goat Polyclonal Antibody (Biotin)
Antibodies
Goat anti-mouse IgG (H&L), F(ab')2 fragment - Affinity Pure, Biotin Conjugate

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR