Anti-IgY Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Tonbo Biosciences
35-8081-M002
76040-272EA 172.7 USD
76040-272
Anti-IgY Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Goat anti-Chicken IgY (H&L) - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: IgY
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR