Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: Tonbo Biosciences
50-8033-U100
76040-232EA 190.3 USD
76040-232
Anti-IgG Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Goat anti-mouse IgG (H+L), secondary antibody, SureLight® R-Phycoerythrin, 100ug

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR