Anti-IgG Rabbit Polyclonal Antibody

Supplier: Tonbo Biosciences
70-8077-M002
76040-218EA 183.7 USD
76040-218
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Rabbit anti-mouse IgG (H&L) - Affinity Pure, min x w/human serum proteins

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Rabbit
Isotype:
Reactivity: mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR