Anti-IgM µRabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Tonbo Biosciences
35-8075-M001
76040-214EA 280.5 USD
76040-214
Anti-IgM µRabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Rabbit anti-Human IgM (mu chain) - Affinity Pure, FITC Conjugate

Type: Secondary
Antigen: Human IgM (µ chain)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: µ chain
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR