Anti-IgG Goat Polyclonal Antibody

Supplier: Tonbo Biosciences
70-8040-M002
76040-172EA 151.8 USD
76040-172
Anti-IgG Goat Polyclonal Antibody
Antibodies
Goat anti-Mouse IgG (H&L) - Affinity Pure, min x w/bovine, horse, human, pig or rabbit serum protein

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR