Anti-IgM Goat Polyclonal Antibody

Supplier: Tonbo Biosciences
70-8030-M002
76040-152EA 165 USD
76040-152
Anti-IgM Goat Polyclonal Antibody
Antibodies
Goat anti-Human IgM (mu chain) - Affinity Pure, min x w/human IgA + IgG

Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR