Anti-IgA Goat Polyclonal Antibody (Biotin)

Supplier: Tonbo Biosciences
30-8117-M001
76040-146EA 233.2 USD
76040-146
Anti-IgA Goat Polyclonal Antibody (Biotin)
Antibodies
Goat anti-Human IgA (alpha chain) - Affinity Pure, Biotin Conjugate, min x w/bovine, mouse, and rabbit serum proteins

Type: Secondary
Antigen: IgA
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: α chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR