3M™ LifeASSURE™ EMC Series Filter Cartridge
Supplier: 3M Healthcare
7010413599
76033-307CS 7940.03 USD
76033-307
3M™ LifeASSURE™ EMC Series Filter Cartridge
Filters Filter Cartridges
LifeASSURE™ EMC Series filter cartridge is a high-efficiency, naturally hydrophilic Nylon 6,6 filter elements designed to meet the exacting requirements of deionized (DI) water, critical parts cleaning, photoresists, and other UHP chemical applications.
  • No IPA pre-wetting and system flushing required
  • Excellent downstream cleanliness out of the package
  • 100% integrity tested
  • Economic alternative to PTFE filter cartridges
Utilizing Advanced Pleat Technology (APT), our LifeASSURE™ EMC Series filter cartridge provides enhanced flow characteristics with minimal pressure drop.
The naturally hydrophilic Nylon 6,6 membrane provides low extractable, long filter life and effective contaminant reduction. LifeASSURE™ EMC filter cartridges are ideally suited for DI water, critical parts cleaning, photoresists, and other UHP chemical applications where superior downstream cleanliness and high efficiency contaminant reduction at 0.1, 0.2, 0.45, 0.65 and 0.80 µm is required.
Applications include DI water, critical parts cleaning and photoresist.
This filter is available in single and multiple length cartridges with 0.1, 0.2, 0.45, 0.65 and 0.80 µm ratings along with various end modifications.