3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 40 Gal. Drawdown Tank w/Connection Kit, 5598409
Supplier: 3M Healthcare
7000029228
76031-646EA 3946.03 USD
76031-646
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 40 Gal. Drawdown Tank w/Connection Kit, 5598409
Tanks Cylindrical Tanks
3M™ commercial reverse osmosis water storage tanks, model 40 gal. Drawdown tank with connection kit is appropriately sized for boiler-based steamer units up to 300000 BTUs.
  • Reverse osmosis 40 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Polyethylene tank liner and fiberglass wrap tank exterior
Tanks can be plumbed together to accommodate larger steamer operations. Fittings are designed to accept plumbing from 3M™ reverse osmosis systems.