3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 10 Gal. Drawdown Tank, 5598407
Supplier: 3M Healthcare
7000050389
76031-644EA 2196.72 USD
76031-644
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 10 Gal. Drawdown Tank, 5598407
Tanks Cylindrical Tanks
3M™ commercial reverse osmosis water storage tanks, model 10 gal. Drawdown tank is designed for use in steamer applications.
  • Reverse osmosis 10 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Includes connection kit
  • Drawdown Tank is a reverse osmosis storage tank that includes a connection kit.