3M™ High Flow Series Triple Manifold Assembly (3XX), 6228405, 3M
Supplier: 3M Healthcare
7000001829
76030-662EA 2390.31 USD
76030-662
3M™ High Flow Series Triple Manifold Assembly (3XX), 6228405, 3M
Manifolds
3M™ High Flow Series Triple Manifold Assembly (3XX) includes a single inlet port for incoming water and a single outlet port for filtered water. This product features ¾" MNPT inlet and outlet water connections.
  • Features ¾" MNPT inlet and outlet water connections
  • Parallel flow
  • Designed for use with 3M™ High Flow Series water filtration cartridges
Filter manifold assemblies provide a vehicle for appropriate water quality to equipment requiring different water characteristics for various applications. It provides filtration and separation solutions for residential, foodservice, science labs, industrial, marine and other applications. These products are innovatively designed for ease of installation and maintenance. Most manifold models include a pressure gauge to monitor differential water pressure.