3M™ Replacement Cartridge OSC Retrofit, Model CS-94C, 5633502
Supplier: 3M Healthcare
7100048258
76030-629CS 397.46 USD
76030-629
3M™ Replacement Cartridge OSC Retrofit, Model CS-94C, 5633502
Filters Filter Cartridges
3M™ water filtration products model CS-94C OCS retrofit replacement cartridge provides consistent quality water for softening system applications by reducing sediment* and chlorine* taste and odor. Ion exchange media and scale* inhibition provide further softening and equipment protection capabilities. This filter cartridge is compatible with most OCS Everpure® foodservice heads.
  • Provides consistent quality water for softening system applications
  • Reduces sediment* and chlorine* taste and odor from incoming water
  • Built-in scale* inhibition as an economical solution to address the potential of scale build-up in hot beverage applications
  • Ion exchange media provides further softening and equipment protection capabilities
  • Compatible with most OCS Everpure® foodservice heads
Our 3M™ water filtration products model CS-94C OCS retrofit replacement cartridge is designed to be effective at reducing sediment* and chlorine* taste and odor, allowing you to feel more confident in the quality of your water
Ion exchange media and scale* inhibition provide further softening and equipment protection capabilities. This filter cartridge is compatible with most OCS Everpure® foodservice heads, and various 3M™ water filtration products OCS retrofit cartridges are available to meet particular water quality concerns. These features together can make this replacement cartridge a great choice for your office softening system needs.
3M™ water filtration products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns. Third-party certifications, such as NSF and WQA, are an advantage provided on many 3M Water Filtration Products. 3M Purification is a known leader in filtration technology recognized for innovation, trust, and quality around the world.
A History of innovation
*As tested and verified by manufacturer's laboratory.
Everpure is a trademark of PENTAIR FILTRATION SOLUTIONS, LLC.
Caution: Please note capacity in specifications is at 11 gpg hardness (180 mg/L hardness).