3M™ High Flow Series Replacement Cartridge, Model HF37, 5632302
Supplier: 3M Healthcare
7100049863
76030-621CS 1354.32 USD
76030-621
3M™ High Flow Series Replacement Cartridge, Model HF37, 5632302
Filters Filter Cartridges
3M™ high flow series replacement cartridge, model HF37 provides consistent quality water by reducing particulate* and chlorine taste and odor from incoming water. The Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench.
  • Provides consistent quality water
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench