3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP25
Supplier: 3M Healthcare
7100049913
76030-587CS 2234.92 USD
76030-587
3M™ Aqua-Pure™ Under Sink Dedicated Faucet Replacement Water Filter Cartridge EP25
Filters Filter Cartridges
3M™ Aqua–Pure™ under sink dedicated faucet replacement water filter cartridge EP25 is compatible with the Everpure® H–104 filter system and is a great choice in water filtration. This cartridge is NSF Certified to reduce particulate, chlorine taste and odor, parasitic protozoan cysts, asbestos, benzene, lead, mercury, atrazine, lindane, p–Dichlorobenzene carbofuran and 2, 4–D reduction.
  • Provides long cartridge life
  • High sediment reduction efficiency
  • Advanced dual stage carbon–block filtration process provides cleaner, clearer better tasting drinking water
  • Sanitary Quick Change (SQC) cartridge design makes cartridge change–out fast and easy
Best Features: 0.6 gpm/2.3 lpm flow rate, 1150 gallon/4353 L capacity, 25 to 125 psi/172 to 862 kPa pressure range, 4.4 to 37.8 °C (40 to 100 °F) temperature range. To maximize system performance and efficiency, regularly scheduled maintenance is required, including replacement of the filter cartridges at the required interval, at the rated capacity, or sooner if a reduction in water flow occurs. Filter life is dependent on actual use and incoming water quality.
Sanitary Quick Change (SQC) design allows you to easily change out and replace the encapsulated media cartridges with a simple quarter turn without touching the spent filter media or contaminated water. These water filter cartridges, working together with other parts, is certified to reduce many contaminants which may be in your drinking water. In many filtration applications, carbon is often the preferred media of choice to absorb certain impurities and one of the most powerful absorbents available. Carbon is highly effective due to the vast surface area contained within a small footprint. One pound of carbon contains a surface area of approximately 100 to 125 acres and can absorb an extremely wide range of substances which may be in drinking water.