3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 5 Gal. Drawdown Tank, 5598406
Supplier: 3M Healthcare
7100079905
76030-242EA 1375.25 USD
76030-242
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 5 Gal. Drawdown Tank, 5598406
Tanks Cylindrical Tanks
3M™ Commercial reverse osmosis water storage tanks, model 5 gal. Drawdown tank is designed for use in steamer applications.
  • Reverse osmosis 5 gallon drawdown tank at 60 psi with 15 psi pre-charge
  • Includes connection kit