3M™ Aqua-Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS24 EPE-316L
Supplier: 3M Healthcare
7000051155
76029-042CS 8580.95 USD
76029-042
3M™ Aqua-Pure™ Whole House Large Diameter Stainless Steel Water Filter Housing SS24 EPE-316L
Filters Filter Cartridges
Large capacity system reduces sediment and other contaminants (cartridge dependent) to help provide cleaner, clearer water from the water used throughout your entire home or facility.
  • Electropolish exterior finish is resistant to corrosion
  • For hot and cold-water applications
  • High flow design
3M™ Aqua–Pure™ SS24 EPE–316L Whole House Filtration System helps provide long lasting, large capacity filtration in harsh corrosive environments. 316L Electropolished Stainless steel. 3" 150# Flange inlet.