3M™ Aqua-Pure™ Whole House Sanitary Quick Change Replacement Water Filter Cartridge AP917HD-S
Supplier: 3M Healthcare
7000029542
76028-694EA 898.72 USD
76028-694
3M™ Aqua-Pure™ Whole House Sanitary Quick Change Replacement Water Filter Cartridge AP917HD-S
Filters Filter Cartridges
3M™ Aqua–Pure™ Whole House Sanitary Quick–Change Replacement Water Filter Cartridge AP917HD–S is suitable for the AP904 system as an activated carbon filter with a nominal particulate reduction rating of 5 microns. This cartridge design is suitable for use on both chlorinated (municipal) and non–chlorinated (well water) system.
  • AP917HD–S Activated carbon filter reduces particulate down to 5–micron nominal and provides scale inhibition
  • Sanitary Quick Change (SQC) design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Up to 10 gallons per minute (GPM) flow rate
  • Whole house water filtration helps reduce sediment build–up in water heaters and appliances
Our 3M™ Aqua–Pure™ Whole House Sanitary Quick–Change Replacement Filter Cartridge AP917HD–S filter reduces sediment and chlorine taste & odor along with providing scale inhibition at a flow rate of up to 10 GPM to meet the needs of larger homes with at least 2.5 bathrooms
We designed the filter to incorporate scale inhibition media to help prevent scale build–up in water–using appliances and piping. Our scale inhibition feature uses polyphosphates in minute quantities to bind calcium and magnesium hardness minerals, preventing them from precipitating out and building up on metallic surface.
Our innovative Sanitary Quick Change (SQC) design, allows for fast and easy cartridge change–outs without having to contact the filter media or the need for a filter wrench. Our Sanitary Quick Change (SQC) is one of the industry's most advanced whole house water filtration system features for this application and minimizes contact with the filter media.
Features include: Filter cartridge is O–ring seal type; Maximum flow rate is up to 20 gpm/75.7 lpm*, AP917HD–S Filter Cartridge incorporates carbon block filtration with a non–woven pleat media plus Cale inhibition, Cartridges are sanitary in design, requiring no contact with the filter media during cartridge change–out.
For more than a century, 3M has applied innovation and technology to improving our customers' lives and supporting their business goals. Today the company, from its headquarters in St. Paul, Minnesota, serves industrial and commercial customers and consumers in nearly 200 countries. The 3M brand, across thousands of products, represents consistency, superior quality, and value. Our success begins with our culture of creative collaboration that inspires a never–ending stream of powerful technologies and product solutions that make life better. We have an ability to apply our technologies – often in combination – to an endless array of real–world customer needs. Of course, all of this made possible by the people of 3M, and their singular commitment to make life easier and better for people around the world. 3M is committed to providing innovative, cost–effective products that help protect people, equipment, and our environment. 3M Purification Inc. (formerly CUNO Incorporated), provides filtration and separation solutions for residential, food service, science labs, industrial, marine, and other applications.
Ordering information: *As tested by manufacturer's laboratory.