3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 2.5 Gal. Drawdown Tank, 5598405
Supplier: 3M Healthcare
7000050388
76028-650EA 597.12 USD
76028-650
3M™ Commercial Reverse Osmosis Water Storage Tanks, Model 2.5 Gal. Drawdown Tank, 5598405
Tanks Cylindrical Tanks
3M™ Commercial reverse osmosis water storage tanks, model 2.5 gal. Drawdown Tank is a reverse osmosis storage tank.
  • Reverse osmosis 2.5 gallon drawdown tank at 60 psi with 15 psi pre-charge
3M™ Water Filtration Products systems and cartridges offer a wide range of solutions designed to address many types of water quality concerns.