3M™ CFS-M RO Membrane for SGLP-075, 5598729
Supplier: 3M Healthcare
7100048971
76028-341CS 3530.78 USD
76028-341
3M™ CFS-M RO Membrane for SGLP-075, 5598729
Filters Filter Cartridges
3M™ CFS–M RO membrane for SGLP–RO has a sanitary quick change (SQC) design allows for easy cartridge change–outs.
  • Provides consistent quality water
  • Reverse osmosis replacement membrane for TDS reduction in boiler less steamers, combi–ovens and flash steamers
  • Sanitary Quick Change (SQC) encapsulated cartridge design allows for fast and easy cartridge change–outs without the need for a filter wrench
  • Replacement RO membrane for model SGLP–RO reverse osmosis systems
It is designed to reduce total dissolved solids (TDS) in boiler less steamers, combi–ovens and flash steamers.