MicroSeal® VP-Sealed Edge Flat Mop Wiper, Berkshire
Supplier: Berkshire
MSVP1018B10
76027-820CS 434.2 USD
76027-820
MicroSeal® VP-Sealed Edge Flat Mop Wiper, Berkshire
Mops
The MicroSeal® VP- Mop Wiper is a sealed edge, laundered, knit wiper.
  • Lightweight
  • Tight knit construction
  • Low abrasion
It is composed of 100% continuous filament polyester knit fabric. This combination of properties provides it with the highest level of cleanliness, abrasion resistance, and chemical compatibility.
The lightweight, tight-knit construction ensures low abrasion, which limits picking and snagging. It is critically low in particles, fibers, ions, and extractables.
Certifications: It is an ultrasonically sealed edge cleanroom laundered wiper required for ISO Class 3 and above environments.