VWR® Upright Freezer Rack for 2" or 3" Boxes
Supplier: VWR International
UF-412L
76027-620EA 98.15 USD
76027-620
VWR® Upright Freezer Rack for 2" or 3" Boxes
Racks Cryogenic Box Racks
Organize Freezer Samples
Racks are designed for use with either standard 2" or 3" cardboard, plastic, or metal boxes. Racks are made from corrosion-resistant stainless steel. Security lock devices are also available.