Accredo Safety Pin And Sleeve - Socket Lockout, ZING Enterprises
Supplier: ZING Enterprises
7264 7265 7266 7267
76026-366EA 42.48 USD
76026-366 76026-368 76026-370 76026-372
Accredo Safety Pin And Sleeve - Socket Lockout, ZING Enterprises
Security Locks
A variety of sizes and configurations are available.
  • High quality
  • Effective lockout device
  • Very durable
Pin and Sleeve Socket Lockout, Small, prevents electrical plugs from being inserted into sockets
Pin and Sleeve Socket Lockout, Medium, prevents electrical plugs from being inserted into sockets. The device is for use with plugs from 220-240V.
Pin and Sleeve Socket Lockout, Large, prevents electrical plugs from being inserted into sockets. The device is for use with plugs from 380-415V.
Extra Large Pin and Sleeve Socket Lockout prevents electrical plugs from being inserted into sockets. The device is for use with 500V plugs.
Certifications: OSHA 1910.147