Accredo Safety Push Button Lockout-Electrical Panels, ZING Enterprises
Supplier: ZING Enterprises
7253
76026-344EA 32.91 USD
76026-344
Accredo Safety Push Button Lockout-Electrical Panels, ZING Enterprises
Security Locks
Supports OSHA Standard 1910.147.
  • Durable Plastic (Polyurethane) Construction
  • High quality
  • Safety device
Push Button Lockout provides a safe and effective method for locking out emergency push buttons. The lockout is made from durable plastic (polyurethane) and measures 2.25’’H x 4’’L x 4’’W. The lockout accommodates 1 padlock with a maximum shackle diameter of ¼’’. Includes a label with the legend, “DANGER Do Not Remove.”
Certifications: OSHA 1910.147