Accredo Safety Stainless Steel Hasp, ZING Enterprises
Supplier: ZING Enterprises
7242
76026-322EA 28.8 USD
76026-322
Accredo Safety Stainless Steel Hasp, ZING Enterprises
Security Locks
Lockout tagout hasp.
  • Supports OSHA Standard 1910.147
  • High quality
  • Very durable
Stainless Steel Hasp is designed to secure switches, valves and other energy devices from use while equipment is being serviced or repaired. The hasp accommodates 7 padlocks. The overall dimensions are as follows: 3.5"L x 2.25"W.
Certifications: OSHA 1910.147