ZING Green Safety RecycLockout Lockout Tagout, Forklift Propane Tank, ZING Enterprises
Supplier: ZING Enterprises
7293
76026-310EA 24.25 USD
76026-310
ZING Green Safety RecycLockout Lockout Tagout, Forklift Propane Tank, ZING Enterprises
Security Locks
Hollow cylindrical shell of 100% recycled polycarbonate that surrounds LP gas cylinder handwheels with a padlock (sold separately).
  • Fits electrical LP gas cylinder handwheels up to 2-3/16
  • Made in USA
  • High quality
ZING RecycLockout Forklift Propane Tank Lockout, locks out LP gas cylinder handwheels up to 2-3/16" diameter, accommodates 3 padlocks with shackles up to 9/16" diameter, 100% recycled yellow HDPE with highly visible label "DO NOT OPERATE", UL Validated, supports sustainability initiatives and LEED credits, measures 2-1/2" Dia. x 3-1/2"W x 3-3/8"H, Made in the USA.
Certifications: 29 CFR 1910.147