ZING Green Safety Stem Shield Valve Protective Cover, ZING Enterprises
Supplier: ZING Enterprises
7211 7212 7217 7218 7219
76026-292EA 136.34 USD
76026-292 76026-294 76026-296 76026-298 76026-300
ZING Green Safety Stem Shield Valve Protective Cover, ZING Enterprises
Security Locks
Available in 5 sizes, covering 95% of all industrial stems.
  • Protects valve from thread damage
  • Ensuring the valve is operational at all times
  • Made in USA
Stem Shield protective valve coverings provide a simple, cost-effective safeguard against injuries sustained when workers bump, graze, or ram into stems that project from industrial valves
Valve Protection: The stems are protected from thread damage and from workplace elements such as corrosion and over sprays. Stem Shield also helps prevent yoke contamination ensuring your valve will operate correctly when the time comes.
Lubricant Protection: The lubrication needed to keep stems operational is protected from the elements and debris, like dirt, which causes wheels to tighten up and become difficult to turn. Lubricant runoff is also eliminated.
Stem Shield covers are lightweight, durable and designed to fit length and diameter of 95% of all industrial stems. Stem Shield is quick and easy to install and does not interfere with valve operation. Stem Shield is constructed utilizing stainless steel attachments and chemically resistant, waterproof, flexible rubber foam cushioning that resist damage, high temperatures, and element erosion. Valve coverings cannot accidently be dislodged and provide long-term safety insurance against worker injury.
Certifications: 29 CFR 1910.36 (g)(1)