ZING Green Safety RecycLockout Breaker Lockout, Universal 3 Pack, ZING Enterprises
Supplier: ZING Enterprises
7110 7111
76026-236PK 58.29 USD
76026-236 76026-238
ZING Green Safety RecycLockout Breaker Lockout, Universal 3 Pack, ZING Enterprises
Security Locks
Circuit Breaker Lockout offered for Single-Pole or Multi-Pole breakers.
  • UL Validated for recycled content
  • Made in USA
  • High quality
Universal Single-Pole Circuit Breaker Lockout is designed with a slide-on feature that allows the device to be securely mounted to a wide range of breakers (110/220 voltage)
Universal Multi-Pole Circuit Breaker Lockout is designed with a slide-on feature that allows the device to be securely mounted to a wide range of breakers (120/277 voltage). Lockout fits double and triple pole switches .3" - .6 " high, .35" - .44" deep, with or without a hole and will accommodate 1 padlock with shackles up to 9/32" diameter. Breaker lockout is high visible red with label "DO NOT OPERATE" and is constructed from heavy-duty 100% recycled polycarbonate that is UL Validated. Recycled materials support sustainability initiatives and LEED credits. Measures 2"L x 1-5/8"W x 1"H. 3/Pack. Made in the USA.
Lockout fits single pole switches .3" - .6 " high, .35" - .44" deep, with or without a hole and will accommodate 1 padlock with shackles up to a 9/32" diameter. Breaker lockout is high visible red with label "DO NOT OPERATE" and is constructed from heavy-duty 100% recycled polycarbonate that is UL Validated. Recycled materials support sustainability initiatives and LEED credits. Measures 2"L x .75"W x 1"H. 3/Pack. Made in the USA.
Certifications: 29 CFR 1910.147