Eco Safety Tag Center, 8 Hook
Supplier: ZING Enterprises
2670
76024-808EA 123.67 USD
76024-808
Eco Safety Tag Center, 8 Hook
Security Locks
Features 8 durable custom hooks that accommodate up to 10 tags per hook.
  • Made in USA
  • High quality
  • Helps organize tags
ZING Eco Safety Tag Center is designed to organize and store a variety of tags commonly used in safety applications such as lockout tagout or equipment inspections. Tag center features 8 durable custom hooks that accommodate up to 10 tags per hook. Station is made from heavy-duty impact resistant recycled PETG that is high visible green to attract attention and encourage use. Recycled materials support sustainability initiatives and LEED credits. Station includes pre-drilled holes for easy mounting (hardware not included) and measures 15"H x 15"W. Made in the USA. (ZING Safety Tags Sold Separately).
Certifications: 29 CFR 1910.147