Frozen-EZ Yeast Transformation II Kit™, Zymo Research
Supplier: Zymo Research
T2001
76020-472KT 160.12 USD
76020-472
Frozen-EZ Yeast Transformation II Kit™, Zymo Research
Nucleic Acid Reagents Transformation Reagents
The Frozen-EZ Yeast Transformation II Kit is designed to make yeast transformations and library screening easier and more efficient than currently available methods.
Yeast cells with high transformation efficiencies can be prepared in under 10 minutes.
Simple method for transforming yeast with single or multiple plasmids in less than 1 hour.
No carrier DNA required.
The yeast cells prepared with the Frozen-EZ Yeast Transformation II Kit™ can be used immediately for transformation or can be stored (i.e., ≤ -70°C) for use at a later time. Yeast prepared with this kit can be transformed with both circular and linear DNAs. Also, the Frozen-EZ Yeast Transformation II Kit™ can be used with other fungi including C. albicans, S. pombe, and P. pastoris.