Quick-DNA/RNA™ Blood Tube Kit, Zymo Research
Supplier: Zymo Research
The Quick-DNA/RNA™ Blood Tube Kit is designed for use with the DNA/RNA Shield™ - Blood Collection Tube (R1150), enabling worry-free sample storage at ambient temperatures. The purification procedure uses Zymo-Spin column technology.
- Superior Yields: Recover DNA and/or total RNA (including small/micro RNAs) without sample loss from the DNA/RNA Shield - Blood Collection Tubes (R1150).
- Streamlined: No reagent removal (no pelleting) - Direct sample processing.
- High-Quality: RNA is ready for all downstream applications including Next-Gen Sequencing, RT-PCR, etc. DNase I Included.
Simply bind the sample onto the Zymo-Spin™ IIICG Column with the aid of reservoirs (vacuum compatible). There is no reagent removal and no pelleting for easy direct whole tube processing. High quality DNA and/or total RNA from 3 ml whole blood is eluted into ≥50 µl of DNase/RNase-free water and is ready for any downstream application including RT-PCR, sequencing, etc
Technical Specifications
Equipment: Microcentrifuge, vortex, and vacuum/vacuum manifold
Purity: DNA and RNA is ready for Next-Gen sequencing, RTPCR, microarray, hybridization, etc. A260/A280, A260/A230: >1.8.
Sample Source: For use with DNA/RNA Shield™ - Blood Collection Tube (prefilled with 6 mL of DNA/RNA Shield™) for the direct collection of up to 3 ml whole-blood (human or animal).
Size Range: Genomic DNA ≥40 kb and Total RNA ≥17 nt
Yield: 50 µg DNA/RNA (binding capacity), ≥ 50 µl (elution volume)
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...