DNA/RNA Shield™ Fecal Collection Tubes

Supplier: Zymo Research
R1101
76020-422EA 148.75 USD
76020-422
DNA/RNA Shield™ Fecal Collection Tubes
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Faeces Sample Collection

The DNA/RNA Shield™ fecal collection tube is powered by the DNA/RNA shield sample stabilization reagent, which takes a microbial snapshot of a sample, inactivates viruses, and makes samples safe and transport ready. Samples are stable at ambient temperature and can be frozen for longer-term storage.



  • Spoon attached to screwcap
  • RNA: Ambient temperature (4 to 25 °C) >1 month
  • DNA: Ambient temperature (4 to 25 °C) >2 years
  • DNA and RNA: Frozen (<–20 °C) indefinitely

Samples collected are ready for all sensitive downstream analyses (microbiomic, next-generation sequencing, qPCR, microarray and more.

DNA/RNA transport medium for stool samples. 1 g or 1 ml stool, soil, environmental samples can be collected.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR