ZR Small-RNA™ PAGE Recovery Kit, Zymo Research
Supplier: Zymo Research
R1070
76020-408EA
168.52
USD
76020-408
ZR Small-RNA™ PAGE Recovery Kit, Zymo Research
Nucleic Acid Reagents
Nucleic Acid Purification Kits and Reagents
The ZR small-RNA™ PAGE Recovery Kit provides an easy and efficient method for the extraction of high quality small RNAs from polyacrylamide gels (native and/or denatured).
- For efficient recovery of small RNA fragments from polyacrylamide gels
- Zymo-Spin™ column technology allows RNA to be eluted into minimal volumes
- Compatible with up to 25% (w/v) polyacrylamide
- RNA is ready for all subsequent analysis and molecular manipulation
The ZR small-RNA™ PAGE Recovery Kit is a refinement of the "crush and soak" method that incorporates a unique buffer system together with Zymo-Spin™ column technologies for improved recovery and added convenience. The recovered RNA can be concentrated into volumes as small as 6 µL, making it ideal for many downstream enzymatic reactions and manipulations.