DNA-OFF™ Cleaning Solution, MP Biomedicals
Supplier: MP BIOMEDICALS (FKA ICN BIOMED
11QD0500
76020-300EA 111.85 USD
76020-300
DNA-OFF™ Cleaning Solution, MP Biomedicals
Cleaning Agents Decontaminants
DNA-OFF™ is a non-alkaline cleaning solution that is highly active against DNA contamination.
  • Ready to use solution
  • Non-carcinogenic
  • Non-corrosive
  • Heat resistant
DNA-OFF™ solution contains a surfactant plus an agent that destroys DNA intended for use at PCR workstations including countertops, apparatus and pipettors
DNA-OFF™ is used for eliminating DNA from all surfaces. It contains a surfactant plus an agent that destroys DNA intended for use at PCR workstations including countertops, apparatus and pipettors. It is stable and heat resistant. In addition, the solution is non-corosive and does not contain carcignogenic compounds.
Presence of contaminant DNA at a PCR work area can result in DNA amplification artifacts. It is stable and heat resistant. In addition, the solution is non-corrosive and does not contain carcinogenic compounds.