IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Warning

Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside

114064112
76020-286EA 281.79 USD
76020-286
IPTG (isopropyl-β-D-thiogalactopyranoside) ≥99%, powder
IPTG (Isopropyl-β-D-thiogalactopyranoside)

  • Storage temperature: -20°C, desiccate

Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 105…115 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now

Specification Test Results

Identity Test Passes
Purity ≥99%
FTIR Conforms to Standard
pH (4% aq) 5.0-7.0
Optical Rotation (c=1, water) -26° to -33°

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR