X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥99% (by TLC), white powder, Ultrapure

Supplier: 0000012207

114063102
76020-284EA 212.02 USD
76020-284
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside) ≥99% (by TLC), white powder, Ultrapure
X-Gal (5-Bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)

  • Ultra Pure Grade
  • Purity: (TLC) >99%
  • Optical Rotation: -61.9%(c=1, DMF:water=1:1, Lit.)
  • Presentation: White Powder
  • Soluble in dimethylformamide (2% in DMF), Dimethyl sulfoxide (20 mg/mL) & DMF:MeOH (1:1) (50 mg/mL)
  • Storage temperature: -20°C

Formula: C₁₄H₁₅BrClNO₆
MDL Number: MFCD00005666
CAS Number: 7240-90-6

Order Now

Specification Test Results

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR